ENAH5802E84D Enamine

(2E)-3-(1-benzofuran-5-yl)prop-2-enoic acid

  • Cas Number : 1210059-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7444943

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7444943
Availability details for
Quantity
  • Availability
  • Add to Cart