ENAH5802E850 Enamine

5-phenylfuran-3-carboxylic acid

  • MDL number MFCD20642175
  • Cas Number : 600158-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1658997

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1658997
Availability details for
Quantity
  • Availability
  • Add to Cart