ENAH5802E85D Enamine

1,1-difluorospiro[2.5]octane-5-carboxylic acid

  • MDL number MFCD31924560
  • Cas Number : 2306264-35-5
  • Molecular Weight : 190.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6498832

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6498832
Availability details for
Quantity
  • Availability
  • Add to Cart