ENAH5802E85E Enamine

tert-butyl 7-amino-decahydroquinoline-1-carboxylate

  • MDL number MFCD24848164
  • Cas Number : 1823260-93-0
  • Molecular Weight : 254.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 7-aminooctahydroquinoline-1(2H)-carboxylate
Inchi Code 1S/C14H26N2O2/c1-14(2,3)18-13(17)16-8-4-5-10-6-7-11(15)9-12(10)16/h10-12H,4-9,15H2,1-3H3
Inchi Key OEUKWLNTAKCQCJ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6839379

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6839379
Availability details for
Quantity
  • Availability
  • Add to Cart