ENAH5802E85F Enamine

methyl 2-amino-3,5,6-trichlorobenzoate

  • MDL number MFCD12159908
  • Cas Number : 1183600-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434378
Availability details for
Quantity
  • Availability
  • Add to Cart