ENAH5802E861 Enamine

7,8-dimethylimidazo[1,2-a]pyridine-2-carboxylic acid hydrochloride

  • MDL number MFCD32065974
  • Cas Number : 2361643-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7431546

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431546
Availability details for
Quantity
  • Availability
  • Add to Cart