ENAH5802E862 Enamine

2-{bicyclo[1.1.1]pentane-1-sulfonyl}acetic acid

  • MDL number MFCD31924232
  • Cas Number : 2344680-35-7
  • Molecular Weight : 190.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(bicyclo[1.1.1]pentan-1-ylsulfonyl)acetic acid
Inchi Code 1S/C7H10O4S/c8-6(9)4-12(10,11)7-1-5(2-7)3-7/h5H,1-4H2,(H,8,9)
Inchi Key QBXKDNVQOKRKFW-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6763072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763072
Availability details for
Quantity
  • Availability
  • Add to Cart