ENAH5802E863 Enamine

5-(1H-1,2,3,4-tetrazol-1-yl)pyridine-3-carboxylic acid

  • MDL number MFCD18276439
  • Cas Number : 1341062-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2690393

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2690393
Availability details for
Quantity
  • Availability
  • Add to Cart