ENAH5802E864 Enamine

5-hydroxy-2-methyl-1H-indole-3-carboxylic acid

  • MDL number MFCD01365791
  • Cas Number : 71982-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187-188
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-143363

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-143363
Availability details for
Quantity
  • Availability
  • Add to Cart