ENAH5802E865 Enamine

1,5-dimethyl-1H-pyrazolo[4,3-b]pyridine-6-carboxylic acid

  • MDL number MFCD31924495
  • Cas Number : 2344679-54-3
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,5-dimethyl-1H-pyrazolo[4,3-b]pyridine-6-carboxylic acid
Inchi Code 1S/C9H9N3O2/c1-5-6(9(13)14)3-8-7(11-5)4-10-12(8)2/h3-4H,1-2H3,(H,13,14)
Inchi Key KBQIRNOCRFAVRT-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7426603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426603
Availability details for
Quantity
  • Availability
  • Add to Cart