ENAH5802E86D Enamine

3,3-dimethyl-2-phenylbutanoic acid

  • MDL number MFCD20483331
  • Cas Number : 13490-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-105
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2156845

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2156845
Availability details for
Quantity
  • Availability
  • Add to Cart