ENAH5802E86F Enamine

2-[4-(methylcarbamoyl)phenyl]acetic acid

  • MDL number MFCD31630676
  • Cas Number : 1269489-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7427715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427715
Availability details for
Quantity
  • Availability
  • Add to Cart