ENAH5802E870 Enamine

2-(pyrrolidin-1-yl)pyrimidine-4-carboxylic acid hydrochloride

  • MDL number MFCD32066591
  • Cas Number : 2361643-61-8
  • Molecular Weight : 229.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(pyrrolidin-1-yl)pyrimidine-4-carboxylic acid hydrochloride
Inchi Code 1S/C9H11N3O2.ClH/c13-8(14)7-3-4-10-9(11-7)12-5-1-2-6-12;/h3-4H,1-2,5-6H2,(H,13,14);1H
Inchi Key GGIIQDNTBSZNPX-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7432702

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7432702
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart