ENAH5802E871 Enamine

4-[(methylcarbamoyl)methyl]benzoic acid

  • MDL number MFCD31924339
  • Cas Number : 2344685-24-9
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(2-(methylamino)-2-oxoethyl)benzoic acid
Inchi Code 1S/C10H11NO3/c1-11-9(12)6-7-2-4-8(5-3-7)10(13)14/h2-5H,6H2,1H3,(H,11,12)(H,13,14)
Inchi Key YPRZYKHZGFQSPK-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7425704
Availability details for
Quantity
  • Availability
  • Add to Cart