ENAH5802E879 Enamine

5-(thiophen-3-yl)furan-3-carboxylic acid

  • MDL number MFCD30309241
  • Cas Number : 1881814-87-4
  • Molecular Weight : 194.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(thiophen-3-yl)furan-3-carboxylic acid
Inchi Code 1S/C9H6O3S/c10-9(11)7-3-8(12-4-7)6-1-2-13-5-6/h1-5H,(H,10,11)
Inchi Key CWLPWAMXFCNXFY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-5200950

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-5200950
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart