ENAH5802E886 Enamine

1-methyl-5-(oxolan-2-yl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD32066339
  • Cas Number : 1888891-92-6
  • Molecular Weight : 196.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7430937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430937
Availability details for
Quantity
  • Availability
  • Add to Cart