ENAH5802E888 Enamine

2,6-dimethoxy-3-methylbenzoic acid

  • MDL number MFCD20639205
  • Cas Number : 96897-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191762

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191762
Availability details for
Quantity
  • Availability
  • Add to Cart