ENAH5802E889 Enamine

3,6-dimethylimidazo[2,1-b][1,3]thiazole-5-carboxylic acid

  • MDL number MFCD11221971
  • Cas Number : 86933-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383122
Availability details for
Quantity
  • Availability
  • Add to Cart