ENAH5802E88B Enamine

5,5-dimethyl-4H,5H,6H-cyclopenta[b]thiophene-2-carboxylic acid

  • MDL number MFCD24502229
  • Cas Number : 1346672-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2835213

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2835213
Availability details for
Quantity
  • Availability
  • Add to Cart