ENAH5802E88C Enamine

2,2-dicyclopentylacetic acid

  • MDL number MFCD20640282
  • Cas Number : 55640-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-60
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7429964

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7429964
Availability details for
Quantity
  • Availability
  • Add to Cart