ENAH5802E88F Enamine

3-chloro-5-cyclopropylbenzoic acid

  • MDL number MFCD19439935
  • Cas Number : 945717-58-8
  • Molecular Weight : 196.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-5-cyclopropylbenzoic acid
Inchi Code 1S/C10H9ClO2/c11-9-4-7(6-1-2-6)3-8(5-9)10(12)13/h3-6H,1-2H2,(H,12,13)
Inchi Key MHVOAKYRJQGWLA-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-124934

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-124934
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart