ENAH5802E895 Enamine

1-(6-oxopiperidin-3-yl)cyclobutane-1-carboxylic acid

  • MDL number MFCD26730859
  • Cas Number : 1552101-65-1
  • Molecular Weight : 197.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(6-oxopiperidin-3-yl)cyclobutane-1-carboxylic acid
Inchi Code 1S/C10H15NO3/c12-8-3-2-7(6-11-8)10(9(13)14)4-1-5-10/h7H,1-6H2,(H,11,12)(H,13,14)
Inchi Key CLJWGPYTRRHFKP-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7354763

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354763
Availability details for
Quantity
  • Availability
  • Add to Cart