ENAH5802E896 Enamine

2-(propan-2-ylsulfanyl)pyridine-3-carboxylic acid

  • MDL number MFCD09727788
  • Cas Number : 152870-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58083

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58083
Availability details for
Quantity
  • Availability
  • Add to Cart