ENAH5802E89B Enamine

3-(dimethylamino)-4,4,4-trifluoro-3-methylbutanoic acid hydrochloride

  • MDL number MFCD32068632
  • Cas Number : 2361736-66-3
  • Molecular Weight : 235.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (S)-3-(dimethylamino)-4,4,4-trifluoro-3-methylbutanoic acid hydrochloride

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7427557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427557
Availability details for
Quantity
  • Availability
  • Add to Cart