ENAH5802E89D Enamine

lithium(1+) 3-(pyridin-4-yl)pyrazine-2-carboxylate

  • MDL number MFCD32065899
  • Cas Number : 2361644-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7430312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430312
Availability details for
Quantity
  • Availability
  • Add to Cart