ENAH5802E8A2 Enamine

3-cyclopropyl-1H-indole-2-carboxylic acid

  • MDL number MFCD32198335
  • Cas Number : 2375273-62-2
  • Molecular Weight : 201.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-cyclopropyl-1H-indole-2-carboxylic acid

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6764589

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764589
Availability details for
Quantity
  • Availability
  • Add to Cart