ENAH5802E8A3 Enamine

3-chloro-5-ethoxypyridine-2-carboxylic acid

  • MDL number MFCD28130301
  • Cas Number : 1425842-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7433532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7433532
Availability details for
Quantity
  • Availability
  • Add to Cart