ENAH5802E8A4 Enamine

8-methoxynaphthalene-1-carboxylic acid

  • MDL number MFCD18412887
  • Cas Number : 5991-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7427654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427654
Availability details for
Quantity
  • Availability
  • Add to Cart