ENAH5802E8A9 Enamine

2-cyclopropyl-1,3-benzoxazole-6-carboxylic acid

  • MDL number MFCD22058022
  • Cas Number : 1369156-25-1
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-cyclopropyl-1,3-benzoxazole-6-carboxylic acid
Inchi Code 1S/C11H9NO3/c13-11(14)7-3-4-8-9(5-7)15-10(12-8)6-1-2-6/h3-6H,1-2H2,(H,13,14)
Inchi Key KJHWEGKKZFHJEP-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7432074

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7432074
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart