ENAH5802E8AB Enamine

2-(1-methyl-1H-1,2,4-triazol-3-yl)benzoic acid

  • Cas Number : 2166780-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5312708

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5312708
Availability details for
Quantity
  • Availability
  • Add to Cart