ENAH5802E8AD Enamine

1-methyl-4-phenyl-1H-1,2,3-triazole-5-carboxylic acid

  • MDL number MFCD23161649
  • Cas Number : 175789-82-9
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-methyl-4-phenyl-1H-1,2,3-triazole-5-carboxylic acid
Inchi Code 1S/C10H9N3O2/c1-13-9(10(14)15)8(11-12-13)7-5-3-2-4-6-7/h2-6H,1H3,(H,14,15)
Inchi Key WDWFXKJOPRBBHR-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-5312689

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-5312689
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart