ENAH5802E8AE Enamine

2-(4-methyl-4H-1,2,4-triazol-3-yl)benzoic acid

  • MDL number MFCD16850794
  • Cas Number : 1247382-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5312701

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5312701
Availability details for
Quantity
  • Availability
  • Add to Cart