ENAH5802E8B1 Enamine

4,4,5,5,5-pentafluoro-3-methylpent-2-enoic acid

  • MDL number MFCD32066635
  • Cas Number : 2361658-52-6
  • Molecular Weight : 204.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (E)-4,4,5,5,5-pentafluoro-3-methylpent-2-enoic acid
Inchi Code 1S/C6H5F5O2/c1-3(2-4(12)13)5(7,8)6(9,10)11/h2H,1H3,(H,12,13)/b3-2+
Inchi Key AIMGIZANJZSIPW-NSCUHMNNSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7429870

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7429870
Availability details for
Quantity
  • Availability
  • Add to Cart