ENAH5802E8B5 Enamine

4-(3-methyl-1H-pyrazol-1-yl)pyrimidine-2-carboxylic acid

  • MDL number MFCD32163411
  • Cas Number : 1781799-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5313073

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5313073
Availability details for
Quantity
  • Availability
  • Add to Cart