ENAH5802E8B9 Enamine

3-(1H-imidazol-4-yl)-2-(1H-pyrrol-1-yl)propanoic acid

  • MDL number MFCD09740805
  • Cas Number : 954584-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-3576698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-3576698
Availability details for
Quantity
  • Availability
  • Add to Cart