ENAH5802E8BA Enamine

3-(1H-1,2,3-benzotriazol-1-yl)-2-methylpropanoic acid

  • MDL number MFCD03043650
  • Cas Number : 4233-62-9
  • Molecular Weight : 205.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 145-146
Iupac Name 3-(1H-1,2,3-benzotriazol-1-yl)-2-methylpropanoic acid
Inchi Code 1S/C10H11N3O2/c1-7(10(14)15)6-13-9-5-3-2-4-8(9)11-12-13/h2-5,7H,6H2,1H3,(H,14,15)
Inchi Key DQDZZJOQSXHOES-UHFFFAOYSA-N

Description

Melting Point 145-146
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6739253

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6739253
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart