ENAH5802E8BE Enamine

7,7-dioxo-7lambda6-thia-1-azabicyclo[3.2.1]octane-5-carboxylic acid

  • MDL number MFCD32066483
  • Cas Number : 2361645-06-7
  • Molecular Weight : 205.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1R,5R)-7-thia-1-azabicyclo[3.2.1]octane-5-carboxylic acid 7,7-dioxide
Inchi Code 1S/C7H11NO4S/c9-6(10)7-2-1-3-8(4-7)13(11,12)5-7/h1-5H2,(H,9,10)
Inchi Key FDIABYBVULQAGP-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7432015

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7432015
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart