ENAH5802E8C3 Enamine

2-(dimethylamino)-1,3-benzoxazole-7-carboxylic acid

  • Cas Number : 1552568-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7425282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7425282
Availability details for
Quantity
  • Availability
  • Add to Cart