ENAH5802E8C4 Enamine

3-benzyloxolane-3-carboxylic acid

  • MDL number MFCD16855839
  • Cas Number : 1493398-27-8
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-benzyltetrahydro-3-furancarboxylic acid
Inchi Code 1S/C12H14O3/c13-11(14)12(6-7-15-9-12)8-10-4-2-1-3-5-10/h1-5H,6-9H2,(H,13,14)
Inchi Key NYPKWCFGOPTYDQ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-5078203
Availability details for
Quantity
  • Availability
  • Add to Cart