ENAH5802E8C7 Enamine

4-bromo-2,2-dimethylpent-4-enoic acid

  • MDL number MFCD24499793
  • Cas Number : 1187330-11-5
  • Molecular Weight : 207.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-2,2-dimethylpent-4-enoic acid
Inchi Code 1S/C7H11BrO2/c1-5(8)4-7(2,3)6(9)10/h1,4H2,2-3H3,(H,9,10)
Inchi Key ACAPMSLCJMXNFT-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6736583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6736583
Availability details for
Quantity
  • Availability
  • Add to Cart