ENAH5802E8CD Enamine

5-methyl-1-(2,2,2-trifluoroethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD20330880
  • Cas Number : 288251-53-6
  • Molecular Weight : 208.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 188-190
Iupac Name 5-methyl-1-(2,2,2-trifluoroethyl)-1H-pyrazole-4-carboxylic acid
Inchi Code 1S/C7H7F3N2O2/c1-4-5(6(13)14)2-11-12(4)3-7(8,9)10/h2H,3H2,1H3,(H,13,14)
Inchi Key YYUGTNNLRIVITH-UHFFFAOYSA-N

Description

Melting Point 188-190
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-5377106

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-5377106
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart