ENAH5802E8CF Enamine

3-(benzyloxy)-3-methylbutanoic acid

  • MDL number MFCD24489928
  • Cas Number : 96556-32-0
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(benzyloxy)-3-methylbutanoic acid
Inchi Code 1S/C12H16O3/c1-12(2,8-11(13)14)15-9-10-6-4-3-5-7-10/h3-7H,8-9H2,1-2H3,(H,13,14)
Inchi Key VNAGEPKLHBWXSX-UHFFFAOYSA-N

Description

Purity 95
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7432833

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart