ENAH5802E8D1 Enamine

2-(1H-imidazol-2-yl)-4-methyl-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD32068740
  • Cas Number : 1881553-17-8
  • Molecular Weight : 209.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(1H-imidazol-2-yl)-4-methylthiazole-5-carboxylic acid
Inchi Code 1S/C8H7N3O2S/c1-4-5(8(12)13)14-7(11-4)6-9-2-3-10-6/h2-3H,1H3,(H,9,10)(H,12,13)
Inchi Key XBVMHGVBNSQROH-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7429839

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7429839
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart