ENAH5802E8D6 Enamine

2-(5-cyclopropyl-1H-pyrazol-1-yl)-3-methoxypropanoic acid

  • MDL number MFCD31811118
  • Cas Number : 2305251-99-2
  • Molecular Weight : 210.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6758954

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6758954
Availability details for
Quantity
  • Availability
  • Add to Cart