ENAH5802E8D7 Enamine

5-(propan-2-yl)-4H,5H,6H,7H-[1,2]oxazolo[4,5-c]pyridine-3-carboxylic acid hydrochloride

  • Cas Number : 2344685-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7427201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427201
Availability details for
Quantity
  • Availability
  • Add to Cart