ENAH5802E8DB Enamine

2-[5-oxo-3-(trifluoromethyl)pyrrolidin-3-yl]acetic acid

  • MDL number MFCD32066553
  • Cas Number : 2361636-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730669

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730669
Availability details for
Quantity
  • Availability
  • Add to Cart