ENAH5802E8DC Enamine

3-fluoro-3-(4-fluorophenyl)cyclobutane-1-carboxylic acid

  • MDL number MFCD31651992
  • Cas Number : 2091608-91-0
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-fluoro-3-(4-fluorophenyl)cyclobutane-1-carboxylic acid
Inchi Code 1S/C11H10F2O2/c12-9-3-1-8(2-4-9)11(13)5-7(6-11)10(14)15/h1-4,7H,5-6H2,(H,14,15)
Inchi Key XLPSAJZYTTYMGS-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-126557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126557
Availability details for
Quantity
  • Availability
  • Add to Cart