ENAH5802E8DF Enamine

4-(3-chlorophenyl)-2-hydroxybut-2-enoic acid

  • Cas Number : 1899087-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432571

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432571
Availability details for
Quantity
  • Availability
  • Add to Cart