ENAH5802E8E1 Enamine

2-(difluoromethyl)-1H-imidazo[4,5-b]pyridine-7-carboxylic acid

  • Cas Number : 1566750-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5408673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5408673
Availability details for
Quantity
  • Availability
  • Add to Cart