ENAH5802E8E2 Enamine

(2S)-2-(hex-5-enamido)-3-methylbutanoic acid

  • Cas Number : 1608762-58-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7439970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7439970
Availability details for
Quantity
  • Availability
  • Add to Cart